1-(1,3-Benzodioxol-5-yl)-1H-pyrrole-2-carbaldehyde 95%
Supplier: Apollo Scientific
OR32345-1G OR32345-5G
APOSOR32345-1GEA 538 EUR
APOSOR32345-1G APOSOR32345-5G
1-(1,3-Benzodioxol-5-yl)-1H-pyrrole-2-carbaldehyde 95%
1-(1,3-Benzodioxol-5-yl)-1H-pyrrole-2-carbaldehyde
Formula: C₁₂H₉NO₃
MW: 215.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03011790
CAS Number: 383147-55-5