Methyl-3-(2-chlorophenyl)-5-methyl-4-isoxazolecarboxylate 95%
Supplier: Apollo Scientific
OR32372-1G OR32372-5G
APOSOR32372-1GEA 336 EUR
APOSOR32372-1G APOSOR32372-5G
Methyl-3-(2-chlorophenyl)-5-methyl-4-isoxazolecarboxylate 95%
Methyl-3-(2-chlorophenyl)-5-methyl-4-isoxazolecarboxylate
Formula: C₁₂H₁₀ClNO₃
MW: 251.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00214711
CAS Number: 4357-94-2