4'-(2-Chloroethyl)acetophenone 95%
Supplier: Apollo Scientific
Synonyms: 1-[4-(2-Chloroethyl)phenyl]ethan-1-one
OR32384-25G OR32384-5G
APOSOR32384-25GEA 887 EUR
APOSOR32384-25G APOSOR32384-5G
4'-(2-Chloroethyl)acetophenone 95%
4'-(2-Chloroethyl)acetophenone
Formula: C₁₀H₁₁ClO
MW: 182.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016350
CAS Number: 69614-95-5