tert-Butyl-4-aminobenzoate
Supplier: Apollo Scientific
|
Warning
|
OR3240-25G
OR3240-5G
OR3240-100G
APOSOR3240-25GEA
24.8
EUR
APOSOR3240-25G
APOSOR3240-5G
APOSOR3240-100G
tert-Butyl-4-aminobenzoate
tert-Butyl 4-aminobenzoate
Formula:
C₁₁H₁₅NO₂ Melting Pt: 108…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00665790 CAS Number: 18144-47-3 |