3,5-Dichlorophenylhydrazinium chloride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3,5-Dichlorophenyl)hydrazine hydrochloride, 3,5-Dichlorophenylhydrazine hydrochloride
OR3249-100G
APOSOR3249-100GEA 130 EUR
APOSOR3249-100G
3,5-Dichlorophenylhydrazinium chloride 98%
3,5-Dichlorophenylhydrazinium chloride
Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
Melting Pt: 204…210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012938
CAS Number: 63352-99-8
EINECS: 264-109-7