2-Chloro-5-methyl[1,6]naphthyridine 95%
Supplier: Apollo Scientific
Synonyms: 2-Chloro-5-methyl-1,6-naphthyridine
OR32650-1G OR32650-250MG
APOSOR32650-1GEA 579 EUR
APOSOR32650-1G APOSOR32650-250MG
2-Chloro-5-methyl[1,6]naphthyridine 95%
2-Chloro-5-methyl[1,6]naphthyridine
Formula: C₉H₇ClN₂
MW: 178.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01443634
CAS Number: 140692-93-9