2,4-Dichloro-5-(2-methoxyethoxy)aniline 95%
Supplier: Apollo Scientific
OR32759-1G OR32759-5G
APOSOR32759-1GEA 538 EUR
APOSOR32759-1G APOSOR32759-5G
2,4-Dichloro-5-(2-methoxyethoxy)aniline 95%
2,4-Dichloro-5-(2-methoxyethoxy)aniline
Formula: C₉H₁₁Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD00974413
CAS Number: 339278-77-2