6-(2,6-Dimethylmorpholino)nicotinaldehyde 95%
Supplier: Apollo Scientific
Synonyms: 6-(2,6-Dimethylmorpholin-4-yl)pyridine-3-carbaldehyde
OR32795-1G OR32795-5G
APOSOR32795-1GEA 306 EUR
APOSOR32795-1G APOSOR32795-5G
6-(2,6-Dimethylmorpholino)nicotinaldehyde 95%
6-(2,6-Dimethylmorpholino)nicotinaldehyde
Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973726
CAS Number: 886360-67-4