2,4-Dichloro-1,3,5-triazine
Supplier: Apollo Scientific
Warning
OR3282-1G OR3282-5G OR3282-25G
APOSOR3282-1GEA 21.9 EUR
APOSOR3282-1G APOSOR3282-5G APOSOR3282-25G
2,4-Dichloro-1,3,5-triazine
2,4-Dichloro-1,3,5-triazine
Formula: C₃HCl₂N₃
Melting Pt: 50…56 °C
Storage Temperature: Freezer
MDL Number: MFCD04115347
CAS Number: 2831-66-5