1-(5-Methyl-4-isoxazolyl)-1-ethanone 95%
Supplier: Apollo Scientific
OR32873-1G OR32873-5G
APOSOR32873-1GEA 336 EUR
APOSOR32873-1G APOSOR32873-5G
1-(5-Methyl-4-isoxazolyl)-1-ethanone 95%
1-(5-Methyl-4-isoxazolyl)-1-ethanone
Formula: C₆H₇NO₂
MW: 125.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01935998
CAS Number: 6497-21-8