2,5-Dimethyl-1-(4H-1,2,4-triazol-4-yl)-1H-pyrrole-3-carbaldehyde 95%
Supplier: Apollo Scientific
OR32900-1G
APOSOR32900-1GEA 180 EUR
APOSOR32900-1G
2,5-Dimethyl-1-(4H-1,2,4-triazol-4-yl)-1H-pyrrole-3-carbaldehyde 95%
2,5-Dimethyl-1-(4H-1,2,4-triazol-4-yl)-1H-pyrrole-3-carbaldehyde
Formula: C₉H₁₀N₄O
Storage Temperature: Ambient
MDL Number: MFCD02187166
CAS Number: 453557-49-8