6-Chloroimidazo[1,2-b]pyridazine 98%
Supplier: Apollo Scientific
Warning
OR3292-25G OR3292-5G OR3292-100G
APOSOR3292-25GEA 78.3 EUR
APOSOR3292-25G APOSOR3292-5G APOSOR3292-100G
6-Chloroimidazo[1,2-b]pyridazine 98%
6-Chloroimidazo[1,2-b]pyridazine
Formula: C₆H₄ClN₃
MW: 153.5691 g/mol
Melting Pt: 115.5…117.5 °C
Storage Temperature: Ambient
MDL Number: MFCD07778345
CAS Number: 6775-78-6