3-(Dimethylamino)propiophenone hydrochloride 95%
Supplier: Apollo Scientific
OR32942-1G OR32942-5G
APOSOR32942-1GEA 26.1 EUR
APOSOR32942-1G APOSOR32942-5G
3-(Dimethylamino)propiophenone hydrochloride 95%
3-(Dimethylamino)propiophenone hydrochloride
Formula: C₁₁H₁₆ClNO
MW: 213.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012481
CAS Number: 879-72-1