4-(4-Methylphenyl)-1,2-dihydrophthalazin-1-one 97%
Supplier: Apollo Scientific
Synonyms: 4-(p-Tolyl)phthalazin-1(2H)-one
OR32943-1G OR32943-5G
APOSOR32943-1GEA 270 EUR
APOSOR32943-1G APOSOR32943-5G
4-(4-Methylphenyl)-1,2-dihydrophthalazin-1-one 97%
4-(4-Methylphenyl)-1,2-dihydrophthalazin-1-one
Formula: C₁₅H₁₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00222976
CAS Number: 51334-85-1