4-(1,3-Benzodioxol-5-yl)benzenecarbaldehyde 95%
Supplier: Apollo Scientific
OR33056-1G OR33056-5G
APOSOR33056-1GEA 336 EUR
APOSOR33056-1G APOSOR33056-5G
4-(1,3-Benzodioxol-5-yl)benzenecarbaldehyde 95%
4-(1,3-Benzodioxol-5-yl)benzenecarbaldehyde
Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782410
CAS Number: 342889-39-8