2-Oxo-4-phenyl-2,5-dihydro-3-furancarbonitrile 95%
Supplier: Apollo Scientific
OR33066-1G
OR33066-25G
OR33066-5G
APOSOR33066-1GEA
180
EUR
APOSOR33066-1G
APOSOR33066-25G
APOSOR33066-5G
2-Oxo-4-phenyl-2,5-dihydro-3-furancarbonitrile 95%
2-Oxo-4-phenyl-2,5-dihydro-3-furancarbonitrile
Formula:
C₁₁H₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00100132 CAS Number: 7692-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...