Imidazo[1,2-a]pyridine-6-boronic acid 98%
Supplier: Apollo Scientific
Warning
OR3308-100MG OR3308-250MG OR3308-1G
APOSOR3308-100MGEA 21.9 EUR
APOSOR3308-100MG APOSOR3308-250MG APOSOR3308-1G
Imidazo[1,2-a]pyridine-6-boronic acid 98%
Imidazo[1,2-a]pyridine-6-boronic acid
Formula: C₇H₇BN₂O₂
MW: 161.9537 g/mol
Melting Pt: 115…117 °C
Storage Temperature: Freezer
MDL Number: MFCD06739225
CAS Number: 913835-63-9