1-(3,4-Dichlorophenyl)-2-(methylsulphonyl)-1-ethanone 95%
Supplier: Apollo Scientific
OR33083-1G OR33083-5G
APOSOR33083-1GEA 270 EUR
APOSOR33083-1G APOSOR33083-5G
1-(3,4-Dichlorophenyl)-2-(methylsulphonyl)-1-ethanone 95%
1-(3,4-Dichlorophenyl)-2-(methylsulphonyl)-1-ethanone
Formula: C₉H₈Cl₂O₃S
MW: 267.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00202208
CAS Number: 344275-49-6