2-Chloro-1-(4-hydroxy-3,5-dimethylphenyl)-1-ethanone 95%
Supplier: Apollo Scientific
OR33090-1G OR33090-5G
APOSOR33090-1GEA 270 EUR
APOSOR33090-1G APOSOR33090-5G
2-Chloro-1-(4-hydroxy-3,5-dimethylphenyl)-1-ethanone 95%
2-Chloro-1-(4-hydroxy-3,5-dimethylphenyl)-1-ethanone
Formula: C₁₀H₁₁ClO₂
MW: 198.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00232525
CAS Number: 40943-25-7