1-Chloro-4-(chloromethylsulphonyl)benzene, Technical Grade
Supplier: Apollo Scientific
Synonyms: p-Chlorophenyl chloromethyl sulphone, (Chloromethyl)(4-chlorophenyl)dioxo-lambda~6~-sulphane
OR33200-1G
APOSOR33200-1GEA 336 EUR
APOSOR33200-1G
1-Chloro-4-(chloromethylsulphonyl)benzene, Technical Grade
(Chloromethyl)(4-chlorophenyl)dioxo-lambda~6~-sulfane
Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00032558
CAS Number: 5943-04-4