(6-Bromoimidazo[1,2-a]pyridin-3-yl)-N,N-dimethylmethanamine hydrochloride 95%
Supplier: Apollo Scientific
OR33201-1G OR33201-5G
APOSOR33201-1GEA 306 EUR
APOSOR33201-1G APOSOR33201-5G
(6-Bromoimidazo[1,2-a]pyridin-3-yl)-N,N-dimethylmethanamine hydrochloride 95%
(6-Bromoimidazo[1,2-a]pyridin-3-yl)-N,N-dimethylmethanamine hydrochloride
Formula: C₁₀H₁₃BrClN₃
MW: 290.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09817424
CAS Number: 1134570-17-4