3-Chloro-N-(2-chloro-3-pyridinyl)-2,2-dimethylpropanamide 95%
Supplier: Apollo Scientific
OR33245-1G OR33245-5G
APOSOR33245-1GEA 336 EUR
APOSOR33245-1G APOSOR33245-5G
3-Chloro-N-(2-chloro-3-pyridinyl)-2,2-dimethylpropanamide 95%
3-Chloro-N-(2-chloro-3-pyridinyl)-2,2-dimethylpropanamide
Formula: C₁₀H₁₂Cl₂N₂O
MW: 247.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00793432
CAS Number: 339100-93-5