3-Hydroxy-2,3-dihydro-4H-furo[3,2-c]chromen-4-one 95%
Supplier: Apollo Scientific
OR33251-1G OR33251-5G
APOSOR33251-1GEA 270 EUR
APOSOR33251-1G APOSOR33251-5G
3-Hydroxy-2,3-dihydro-4H-furo[3,2-c]chromen-4-one 95%
3-Hydroxy-2,3-dihydro-4H-furo[3,2-c]chromen-4-one
Formula: C₁₁H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD01443897
CAS Number: 182115-55-5