2-Azaspiro[4.4]nonan-3-one 95%
Supplier: Apollo Scientific
OR33313-1G
OR33313-5G
APOSOR33313-1GEA
121
EUR
APOSOR33313-1G
APOSOR33313-5G
2-Azaspiro[4.4]nonan-3-one 95%
2-Azaspiro[4.4]nonan-3-one
Formula:
C₈H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD09864498 CAS Number: 75751-72-3 |