4-(4-Methylphenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile 95%
Supplier: Apollo Scientific
OR33315-1G OR33315-5G
APOSOR33315-1GEA 336 EUR
APOSOR33315-1G APOSOR33315-5G
4-(4-Methylphenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile 95%
4-(4-Methylphenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Formula: C₁₃H₁₀N₂O
MW: 210.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056675
CAS Number: 886362-01-2