6-Chloro-2,3-dihydro-4H-thiochromen-4-one oxime 95%
Supplier: Apollo Scientific
Synonyms: N-[(4E)-6-Chloro-3,4-dihydro-2H-1-benzothiopyran-4-ylidene]hydroxylamine
OR33381-1G OR33381-5G
APOSOR33381-1GEA 336 EUR
APOSOR33381-1G APOSOR33381-5G
6-Chloro-2,3-dihydro-4H-thiochromen-4-one oxime 95%
6-Chloro-2,3-dihydro-4H-thiochromen-4-one oxime
Formula: C₉H₈ClNOS
MW: 213.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056679
CAS Number: 1029134-50-6