1-(7-Bromo-5-methoxy-1-benzofuran-2-yl)-1-ethanone 95%
Supplier: Apollo Scientific
OR33390-1G OR33390-5G
APOSOR33390-1GEA 244 EUR
APOSOR33390-1G APOSOR33390-5G
1-(7-Bromo-5-methoxy-1-benzofuran-2-yl)-1-ethanone 95%
1-(7-Bromo-5-methoxy-1-benzofuran-2-yl)-1-ethanone
Formula: C₁₁H₉BrO₃
MW: 269.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02082020
CAS Number: 454473-82-6