2'-(Methylsulphinyl)[1,1'-biphenyl]-4-carbaldehyde 95%
Supplier: Apollo Scientific
OR33473-1G OR33473-5G
APOSOR33473-1GEA 336 EUR
APOSOR33473-1G APOSOR33473-5G
2'-(Methylsulphinyl)[1,1'-biphenyl]-4-carbaldehyde 95%
2'-(Methylsulphinyl)[1,1'-biphenyl]-4-carbaldehyde
Formula: C₁₄H₁₂O₂S
MW: 244.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972222
CAS Number: 1171155-44-4