1-Thia-4-azaspiro[4.5]decan-3-one 95%
Supplier: Apollo Scientific
OR33497-1G OR33497-5G
APOSOR33497-1GEA 270 EUR
APOSOR33497-1G APOSOR33497-5G
1-Thia-4-azaspiro[4.5]decan-3-one 95%
1-Thia-4-azaspiro[4.5]decan-3-one
Formula: C₈H₁₃NOS
MW: 171.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00101403
CAS Number: 4580-63-6