1-Thia-4-azaspiro[4.5]decane hydrochloride 95%
Supplier: Apollo Scientific
OR33498-1G OR33498-5G
APOSOR33498-1GEA 270 EUR
APOSOR33498-1G APOSOR33498-5G
1-Thia-4-azaspiro[4.5]decane hydrochloride 95%
1-Thia-4-azaspiro[4.5]decane hydrochloride
Formula: C₈H₁₆ClNS
MW: 193.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14584815
CAS Number: 933-41-5