3-Oxo-3-[4-(1-pyrrolidinyl)phenyl]propanenitrile 95%
Supplier: Apollo Scientific
OR33526-1G OR33526-5G
APOSOR33526-1GEA 336 EUR
APOSOR33526-1G APOSOR33526-5G
3-Oxo-3-[4-(1-pyrrolidinyl)phenyl]propanenitrile 95%
3-Oxo-3-[4-(1-pyrrolidinyl)phenyl]propanenitrile
Formula: C₁₃H₁₄N₂O
MW: 214.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07786223
CAS Number: 887595-01-9