4-Methyl-1,5,6,7-tetrahydro-2H-cyclopenta[b]pyridin-2-one 95%
Supplier: Apollo Scientific
OR33539-1G OR33539-5G
APOSOR33539-1GEA 270 EUR
APOSOR33539-1G APOSOR33539-5G
4-Methyl-1,5,6,7-tetrahydro-2H-cyclopenta[b]pyridin-2-one 95%
4-Methyl-1,5,6,7-tetrahydro-2H-cyclopenta[b]pyridin-2-one
Formula: C₉H₁₁NO
MW: 149.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16140309
CAS Number: 20594-30-3