5-Bromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5-Bromomeldrum's acid) 95%
Supplier: Apollo Scientific
Synonyms: 5-Bromo-2,2-dimethyl-1,3-dioxane-4,6-dione
OR33544-1G OR33544-5G
APOSOR33544-1GEA 180 EUR
APOSOR33544-1G APOSOR33544-5G
5-Bromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5-Bromomeldrum's acid) 95%
5-Bromo-2,2-dimethyl-1,3-dioxane-4,6-dione (5-Bromomeldrum's acid)
Formula: C₆H₇BrO₄
Storage Temperature: Ambient
CAS Number: 66145-20-8