2-(Phenylthio)ethanamine, Technical Grade
Supplier: Apollo Scientific
Synonyms: 2-(Phenylsulphanyl)ethan-1-amine
OR33589-1G
APOSOR33589-1GEA 270 EUR
APOSOR33589-1G
2-(Phenylthio)ethanamine, Technical Grade
2-(Phenylthio)ethanamine
Formula: C₈H₁₁NS
MW: 153.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014824
CAS Number: 2014-75-7