2,3-Dihydro-2-methylbenzofuran 95%
Supplier: Apollo Scientific
Synonyms: 2-Methyl-2,3-dihydrobenzofuran
OR33590-1G OR33590-5G
APOSOR33590-1GEA 53.8 EUR
APOSOR33590-1G APOSOR33590-5G
2,3-Dihydro-2-methylbenzofuran 95%
2,3-Dihydro-2-methylbenzofuran
Formula: C₉H₁₀O
MW: 134.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023003
CAS Number: 1746-11-8