[4-(tert-Butylsulphanyl)phenyl]methanol 95%
Supplier: Apollo Scientific
OR33595-1G OR33595-5G
APOSOR33595-1GEA 538 EUR
APOSOR33595-1G APOSOR33595-5G
[4-(tert-Butylsulphanyl)phenyl]methanol 95%
[4-(tert-Butylsulphanyl)phenyl]methanol
Formula: C₁₁H₁₆OS
MW: 196.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18890812
CAS Number: 500530-69-8