3-N-Cyclohexylpyridine-2,3-diamine 97%
Supplier: Apollo Scientific
OR33600-1G OR33600-5G
APOSOR33600-1GEA 180 EUR
APOSOR33600-1G APOSOR33600-5G
3-N-Cyclohexylpyridine-2,3-diamine 97%
3-N-Cyclohexylpyridine-2,3-diamine
Formula: C₁₁H₁₇N₃
MW: 191.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20688652
CAS Number: 1286273-78-6