4-Bromo-2-nitrophenol 98%
Supplier: Apollo Scientific
Danger
OR3371-25G OR3371-5G
APOSOR3371-25GEA 45.1 EUR
APOSOR3371-25G APOSOR3371-5G
4-Bromo-2-nitrophenol 98%
4-Bromo-2-nitrophenol
Formula: C₆H₄BrNO₃
MW: 218.01 g/mol
Boiling Pt: 259
Melting Pt: 88…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00082540
CAS Number: 7693-52-9
EINECS: 231-707-4
UN: 2811
ADR: 6.1 III