1-(Pyridin-2-ylmethyl)piperazine 97%
Supplier: Apollo Scientific
Warning
OR3379-1G OR3379-5G
APOSOR3379-1GEA 0 EUR
APOSOR3379-1G APOSOR3379-5G
1-(Pyridin-2-ylmethyl)piperazine 97%
1-(Pyridin-2-ylmethyl)piperazine
Formula: C₁₀H₁₅N₃
MW: 177.25 g/mol
Melting Pt: 91…93/0.4mm °C
Storage Temperature: Ambient
MDL Number: MFCD01862551
CAS Number: 55579-01-6