3-(2-Methoxyphenyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR34-1G OR34-100MG
APOSOR34-1GEA 87 EUR
APOSOR34-1G APOSOR34-100MG
3-(2-Methoxyphenyl)-1H-pyrazole
3-(2-Methoxyphenyl)-1H-pyrazole
Formula: C₁₀H₁₀N₂O
MW: 174.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02091524
CAS Number: 59843-63-9