2-Methoxy-4-nitrobenzonitrile 99%
Supplier: Apollo Scientific
Warning
OR345004-1G OR345004-5G
APOSOR345004-1GEA 50.9 EUR
APOSOR345004-1G APOSOR345004-5G
2-Methoxy-4-nitrobenzonitrile 99%
2-Methoxy-4-nitrobenzonitrile
Formula: C₈H₆N₂O₃
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD02093781
CAS Number: 101084-96-2