4-Methoxy-o-toluic acid
Supplier: Apollo Scientific
Warning
OR345161-25G OR345161-5G
APOSOR345161-25GEA 49.3 EUR
APOSOR345161-25G APOSOR345161-5G
4-Methoxy-o-toluic acid
4-Methoxy-o-toluic acid
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 177…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00020291
CAS Number: 6245-57-4