Imidazo[1,2-a]pyridine-5-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR345351-1G
OR345351-5G
APOSOR345351-1GEA
143
EUR
APOSOR345351-1G
APOSOR345351-5G
Imidazo[1,2-a]pyridine-5-carboxylic acid
Imidazo[1,2-a]pyridine-5-carboxylic acid
Formula:
C₈H₆N₂O₂ MW: 162.1500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09834780 CAS Number: 479028-72-3 |