2-Bromo-5-hydroxy-3-methylpyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-5-hydroxy-3-picoline
OR345364-5G
APOSOR345364-5GEA 116 EUR
APOSOR345364-5G
2-Bromo-5-hydroxy-3-methylpyridine 98%
2-Bromo-5-hydroxy-3-methylpyridine
Formula: C₆H₆BrNO
MW: 188.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839267
CAS Number: 1003711-43-0