4-Bromo-3-iodopyridine
Supplier: Apollo Scientific
Warning
OR345446-1G OR345446-5G
APOSOR345446-1GEA 42.2 EUR
APOSOR345446-1G APOSOR345446-5G
4-Bromo-3-iodopyridine
4-Bromo-3-iodopyridine
Formula: C₅H₃BrIN
MW: 283.8900 g/mol
Melting Pt: 77…78 °C
Storage Temperature: Freezer
MDL Number: MFCD04112578
CAS Number: 89167-20-4