N'-[1-Amino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
OR346001-1G
OR346001-500MG
APOSOR346001-1GEA
0
EUR
APOSOR346001-1G
APOSOR346001-500MG
N'-[1-Amino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formula:
C₁₃H₁₉N₃O₂ MW: 249.32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568176 CAS Number: 159016-22-5 |