1-tert-Butyloxycarbonyl-4-(3-aminopropyl)piperazine 95+%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 4-(3-aminopropyl)piperazine-1-carboxylate
OR346019-5G
APOSOR346019-5GEA 215 EUR
APOSOR346019-5G
1-tert-Butyloxycarbonyl-4-(3-aminopropyl)piperazine 95+%
1-tert-Butyloxycarbonyl-4-(3-aminopropyl)piperazine
Formula: C₁₂H₂₅N₃O₂
MW: 243.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06808578
CAS Number: 373608-48-1