N'-[1-Cyclopropylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
OR346033-1G OR346033-500MG
APOSOR346033-1GEA 0 EUR
APOSOR346033-1G APOSOR346033-500MG
N'-[1-Cyclopropylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Cyclopropylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
Formula: C₁₁H₂₁N₃O₂
MW: 227.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568246
CAS Number: 1053657-24-1