N'-[1-[(Furan-2-ylmethyl)amino]2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
OR346034-1G OR346034-500MG
APOSOR346034-1GEA 0 EUR
APOSOR346034-1G APOSOR346034-500MG
N'-[1-[(Furan-2-ylmethyl)amino]2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-[(Furan-2-ylmethyl)amino]2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formula: C₁₈H₂₃N₃O₃
MW: 329.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568247
CAS Number: 1053657-29-6