N'-[1-Cyclopropylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
OR346036-1G OR346036-500MG
APOSOR346036-1GEA 0 EUR
APOSOR346036-1G APOSOR346036-500MG
N'-[1-Cyclopropylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Cyclopropylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formula: C₁₆H₂₃N₃O₃
MW: 305.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568249
CAS Number: 1053657-38-7